Anti-TCR Va24-Ja18 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 6B11]

Supplier: Biolegend
342917 342918
CA10758-668EA 307.99 CAD
CA10758-668 CA10758-670
Anti-TCR Va24-Ja18 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 6B11]
Antibodies
Brilliant Violet 510™ anti-human TCR Vα24-Jα18 (iNKT cell) [6B11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactivity: Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR Va24-Ja18
Clonality: Monoclonal
Clone: 6B11
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR