Anti-TCR gamma/delta Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: B1]

Supplier: Biolegend
331219 331220
CA10758-450EA 325.54 CAD
CA10758-450 CA10758-452
Anti-TCR gamma/delta Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: B1]
Antibodies
Brilliant Violet 510™ anti-human TCR γ/δ [B1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR gamma/delta
Clonality: Monoclonal
Clone: B1
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR