Anti-Siglec H Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 551]

Supplier: Biolegend
129609 129610
CA129609-BLEA 244.16 CAD
CA129609-BL CA129610-BL
Anti-Siglec H Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 551]
Antibodies
Pacific Blue™ anti-mouse Siglec H [551]; Isotype: Rat IgG1; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Siglec H
Clonality: Monoclonal
Clone: 551
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR