Anti-Ovalbumin Mouse Monoclonal Antibody [clone: TOSGAA1]

Supplier: Biolegend
520401 520402
CA10761-908EA 132.45 CAD
CA10761-908 CA10761-910
Anti-Ovalbumin Mouse Monoclonal Antibody [clone: TOSGAA1]
Antibodies
Purified anti-chicken Ovalbumin [TOSGAA1]; Isotype: Mouse IgG2a; Reactivity: Chicken; Apps: ELISA; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA standard, a concentration range of 9.3-600 pg/ml is recommended. It is recommended that the reagent be titrated for optimal performance.

Type: Primary
Antigen: Ovalbumin
Clonality: Monoclonal
Clone: TOSGAA1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR