Anti-NK-1.1 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: PK136]

Supplier: Biolegend
108737 108738
CA10756-892EA 307.99 CAD
CA10756-892 CA10756-894
Anti-NK-1.1 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: PK136]
Antibodies
Brilliant Violet 510™ anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: NK-1.1
Clonality: Monoclonal
Clone: PK136
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR