Anti-NK-1.1 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: PK136]

Supplier: Biolegend
108735 108736
CA108735-BLEA 378.2 CAD
CA108735-BL CA10756-890
Anti-NK-1.1 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: PK136]
Antibodies
Brilliant Violet 650™ anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For flow cytometric staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Order Now

SPECIFICATIONS

Antigen symbol NK-1.1 NK-1.1
Environmentally Preferable
Antigen synonyms NKR-P1C|NKR-P1B|CD161c|Ly-55|NK1.1|KLRB1C|CD161b NKR-P1C|NKR-P1B|CD161c|Ly-55|NK1.1|KLRB1C|CD161b
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation Brilliant Violet® 650 Brilliant Violet® 650
Clone PK136 PK136
Reactivity Mouse Mouse
Host Mouse Mouse
Gene ID 17059 17059
Isotype IgG2a kappa IgG2a kappa
Immunogen NK-1+ cells from mouse spleen and bone marrow NK-1+ cells from mouse spleen and bone marrow
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA). Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 650™ and unconjugated antibody. The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 650™ and unconjugated antibody.
Concentration 0.20 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze. The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR