Anti-NFATc1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 7A6]

Supplier: Biolegend
649603 649604
CA10763-286EA 234.58 CAD
CA10763-286 CA10763-288
Anti-NFATc1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 7A6]
Antibodies
Alexa Fluor® 488 anti-NFATc1 [7A6]; Isotype: Mouse IgG1; Reactivity: Human, Mouse, Rat; Apps: ICFC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: NFATc1
Clonality: Monoclonal
Clone: 7A6
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR