Anti-Nestin Mouse Monoclonal Antibody [clone: 10C2]

Supplier: Biolegend
656801 656802
CA10763-650EA 178.73 CAD
CA10763-650 CA10763-652
Anti-Nestin Mouse Monoclonal Antibody [clone: 10C2]
Antibodies
Purified anti-Nestin [10C2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml.  For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Nestin
Clonality: Monoclonal
Clone: 10C2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR