Anti-MRP-14 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRP 1H9]

Supplier: Biolegend
350705 350706
CA10758-984EA 225.01 CAD
CA10758-984 CA10758-986
Anti-MRP-14 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRP 1H9]
Antibodies
PE anti-human MRP-14 (S100A9) [MRP 1H9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MRP-14
Clonality: Monoclonal
Clone: MRP 1H9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR