Anti-MR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 26.5]

Supplier: Biolegend
361105 361106
CA10760-832EA 244.16 CAD
CA10760-832 CA10760-834
Anti-MR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 26.5]
Antibodies
PE anti-human/mouse/rat MR1 [26.5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat, Cross-Reactivity: Bovine; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MR1
Clonality: Monoclonal
Clone: 26.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Mouse;Rat;Cross-Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR