Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: RB6-8C5]

Supplier: Biolegend
108451 108452
CA10758-868EA 225.01 CAD
CA10758-868 CA10758-870
Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: RB6-8C5]
Antibodies
PE/Dazzle™ 594 anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G/Ly-6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR