Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: RB6-8C5]

Supplier: Biolegend
108437 108438
CA10759-186EA 280.86 CAD
CA10759-186 CA10759-188
Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: RB6-8C5]
Antibodies
Brilliant Violet 510™ anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G/Ly-6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR