Anti-Ly-6C Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: HK1.4]

Supplier: Biolegend
128031 128032
CA10757-098EA 280.86 CAD
CA10757-098 CA10757-100
Anti-Ly-6C Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: HK1.4]
Antibodies
Brilliant Violet 421™ anti-mouse Ly-6C [HK1.4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Brilliant Violet 421™ excites at 405 nm and emits at 421 nm. The standard bandpass filter 450/50 nm is recommended for detection. Brilliant Violet 421™ is a trademark of Sirigen Group Ltd.

Type: Primary
Antigen: Ly-6C
Clonality: Monoclonal
Clone: HK1.4
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2c, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR