Anti-KLRG1 Syrian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: 2F1/KLRG1]

Supplier: Biolegend
138417 138418
CA10757-188EA 197.88 CAD
CA10757-188 CA10757-190
Anti-KLRG1 Syrian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: 2F1/KLRG1]
Antibodies
PerCP/Cy5.5 anti-mouse/human KLRG1 (MAFA) [2F1/KLRG1]; Isotype: Syrian Hamster IgG; Reactivity: Mouse, Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: KLRG1
Clonality: Monoclonal
Clone: 2F1/KLRG1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Syrian Hamster
Isotype: Syrian Hamster IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR