Anti-IRF8 Mouse Monoclonal Antibody [clone: 7G11A45]

Supplier: Biolegend
656501 656502
CA10763-542EA 178.73 CAD
CA10763-542 CA10763-544
Anti-IRF8 Mouse Monoclonal Antibody [clone: 7G11A45]
Antibodies
Purified anti-IRF8 [7G11A45]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF, IP; Size: 25 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. For immunoprecipitation, the suggested use of this reagent is 5-20 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IRF8
Clonality: Monoclonal
Clone: 7G11A45
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR