Anti-IRF4 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: IRF4.3E4]

Supplier: Biolegend
646407 646408
CA10763-274EA 197.88 CAD
CA10763-274 CA10763-276
Anti-IRF4 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: IRF4.3E4]
Antibodies
Alexa Fluor® 647 anti-IRF4 [IRF4.3E4]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IRF4
Clonality: Monoclonal
Clone: IRF4.3E4
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR