Anti-IL-4 Rat Monoclonal Antibody [clone: 11B11]

Supplier: Biolegend
504121 504122
CA504121-BLEA 188.3 CAD
CA504121-BL CA504122-BL
Anti-IL-4 Rat Monoclonal Antibody [clone: 11B11]
Antibodies
Ultra-LEAF™ Purified anti-mouse IL-4 [11B11]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, CyTOF®, ELISPOT Capture, Neut, ICC, IP, IHC; Size: 1 mg

Each lot of this antibody is quality control tested by ELISA assay. For ELISPOT applications, a concentration range of 2-6 μg/ml is recommended. For ELISA capture applications, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-4 recombinant protein ranging from 250 to 2 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: 11B11
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR