Anti-IL-34 Mouse Monoclonal Antibody (Biotin) [clone: E033B8]

Supplier: Biolegend
361401 361402
CA10760-952EA 280.86 CAD
CA10760-952 CA10760-954
Anti-IL-34 Mouse Monoclonal Antibody (Biotin) [clone: E033B8]
Antibodies
Biotin anti-human IL-34 [E033B8]; Isotype: Mouse IgG2a, λ; Reactivity: Human; Apps: ELISA Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.5 - 2.0 μg/mL is recommended. To obtain a linear standard curve, serial dilutions of human IL-34 recombinant protein ranging from 5000 to 78 pg/mL are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-34
Clonality: Monoclonal
Clone: E033B8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, λ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR