Anti-IL-32 Mouse Monoclonal Antibody [clone: KU32-07]

Supplier: Biolegend
513301 513302
CA10761-678EA 217.03 CAD
CA10761-678 CA10761-680
Anti-IL-32 Mouse Monoclonal Antibody [clone: KU32-07]
Antibodies
Purified anti-human IL-32α [KU32-07]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA capture antibody, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of human IL-32 recombinant protein ranging from 500 to 7.8 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-32
Clonality: Monoclonal
Clone: KU32-07
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR