Anti-IL-27 Mouse Monoclonal Antibody [clone: MM27-7B1]

Supplier: Biolegend
516901 516902
CA10761-732EA 178.73 CAD
CA10761-732 CA10761-734
Anti-IL-27 Mouse Monoclonal Antibody [clone: MM27-7B1]
Antibodies
Purified anti-mouse IL-27 p28 [MM27-7B1]; Isotype: Mouse IgG2a; Reactivity: Mouse; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. A concentration of 0.5 μg/ml of the capture antibody was utilized to generate the example standard curve. It is recommended that the end-user titrate each lot of reagents for optimal performance.

Type: Primary
Antigen: IL-27
Clonality: Monoclonal
Clone: MM27-7B1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR