Anti-IL-27 Rat Monoclonal Antibody (Biotin) [clone: B02P6E6]

Supplier: Biolegend
525903 525904
CA10761-796EA 234.58 CAD
CA10761-796 CA10761-798
Anti-IL-27 Rat Monoclonal Antibody (Biotin) [clone: B02P6E6]
Antibodies
Biotin anti-mouse IL-27 [B02P6E6]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ELISA Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. A concentration of 0.125 μg/ml of the capture antibody was utilized to generate the example standard curve. It is recommended that the end-user titrate each lot of reagents for optimal performance.

Type: Primary
Antigen: IL-27
Clonality: Monoclonal
Clone: B02P6E6
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR