Anti-IL-2 Rat Monoclonal Antibody [clone: JES6-1A12]

Supplier: Biolegend
503705 503706
CA503705-BLEA 175.54 CAD
CA503705-BL CA503706-BL
Anti-IL-2 Rat Monoclonal Antibody [clone: JES6-1A12]
Antibodies
Ultra-LEAF™ Purified anti-mouse IL-2 [JES6-1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ELISA Capture, ELISPOT Capture, IP, Neut, WB; Size: 1 mg

Each lot of this antibody is quality control tested by ELISA assay. For ELISPOT capture applications, a concentration range of 2-6 μg/ml is recommended. For ELISA capture applications, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-2 recombinant protein ranging from 250 to 2 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: JES6-1A12
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR