Anti-IL-17A Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: BL168]

Supplier: Biolegend
512323 512324
CA512323-BLEA 395.76 CAD
CA512323-BL CA10761-972
Anti-IL-17A Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: BL168]
Antibodies
Brilliant Violet 570™ anti-human IL-17A [BL168]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-17A
Clonality: Monoclonal
Clone: BL168
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR