Anti-IL-15 Mouse Monoclonal Antibody (Biotin) [clone: BH1543]

Supplier: Biolegend
515103 515104
CA10761-700EA 197.88 CAD
CA10761-700 CA10761-976
Anti-IL-15 Mouse Monoclonal Antibody (Biotin) [clone: BH1543]
Antibodies
Biotin anti-human IL-15 [BH1543]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-15 recombinant protein ranging from 500 to 7.8 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-15
Clonality: Monoclonal
Clone: BH1543
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR