Anti-Ikaros Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 2A9/Ikaros]

Supplier: Biolegend
653305 653306
CA10762-778EA 272.88 CAD
CA10762-778 CA10762-780
Anti-Ikaros Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 2A9/Ikaros]
Antibodies
Alexa Fluor® 647 anti-mouse Ikaros [2A9/Ikaros]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 0.1 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ikaros
Clonality: Monoclonal
Clone: 2A9/Ikaros
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR