Anti-IgM Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: MHM-88]

Supplier: Biolegend
314523 314524
CA10758-136EA 343.1 CAD
CA10758-136 CA10758-138
Anti-IgM Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: MHM-88]
Antibodies
Brilliant Violet 605™ anti-human IgM [MHM-88]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone: MHM-88
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

SPECIFICATIONS

Antigen symbol IgM
Environmentally Preferable
Antigen synonyms Immunoglobulin M|IGHM
Antibody type Primary
Clonality Monoclonal
Conjugation Brilliant Violet® 605
Clone MHM-88
Reactivity Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Human
Host Mouse
Gene ID 3507
Isotype IgG1 kappa
Immunogen Human Ig cocktail
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 605™ and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR