Anti-IgG1, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: RTK2071]

Supplier: Biolegend
400437 400438
CA10761-148EA 325.54 CAD
CA10761-148 CA10761-150
Anti-IgG1, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: RTK2071]
Antibodies
Brilliant Violet 650™ Rat IgG1, κ Isotype Ctrl [RTK2071]; Isotype: Rat IgG1, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2071
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Rat
Order Now

SPECIFICATIONS

Antigen symbol IgG1 k isotype ctrl IgG1 k isotype ctrl
Environmentally Preferable
Antibody type Control Control
Clonality Monoclonal Monoclonal
Conjugation Brilliant Violet® 650 Brilliant Violet® 650
Clone RTK2071 RTK2071
Reactivity Rat Rat
Host Rat Rat
Isotype IgG1 kappa IgG1 kappa
Epitope kappa-chain kappa-chain
Immunogen Trinitrophenol + KLH Trinitrophenol + KLH
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA). Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 650™ and unconjugated antibody. The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 650™ and unconjugated antibody.
Concentration 0.20 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze. The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR