Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: MOPC-21]

Supplier: Biolegend
400169 400170
CA400169-BLEA 325.54 CAD
CA400169-BL CA10761-080
Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: MOPC-21]
Antibodies
Brilliant Violet 785™ Mouse IgG1, κ Isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR