Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody [clone: MOPC-21]

Supplier: Biolegend
400165 400166
CA400165-BLEA 167.56 CAD
CA400165-BL CA400166-BL
Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody [clone: MOPC-21]
Antibodies
Ultra-LEAF™ Purified Mouse IgG1, κ Isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, ICFC, WB, IP, ICC, IF, FA; Size: 1 mg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-21
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR