Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody [clone: MG1-45]

Supplier: Biolegend
401407 401408
CA401407-BLEA 167.56 CAD
CA401407-BL CA401408-BL
Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody [clone: MG1-45]
Antibodies
Ultra-LEAF™ Purified Mouse IgG1, κ Isotype Ctrl [MG1-45]; Isotype: Mouse (BALB/c) IgG1, κ; Reactivity: Mouse; Apps: FC, ICFC, WB, IP, ICC, IF, FA; Size: 1 mg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control, and the purity is greater than 95% by SDS-PAGE. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: MG1-45
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR