Anti-IgG1 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RMG1-1]

Supplier: Biolegend
406615 406616
CA10761-386EA 280.86 CAD
CA10761-386 CA10761-388
Anti-IgG1 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RMG1-1]
Antibodies
Brilliant Violet 421™ anti-mouse IgG1 [RMG1-1]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: RMG1-1
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol IgG1
Environmentally Preferable
Antigen synonyms IGHG1|Immunoglobulin G1
Antibody type Primary
Clonality Monoclonal
Conjugation Brilliant Violet® 421
Clone RMG1-1
Reactivity Mouse
Host Rat
Gene ID 16017
Isotype IgG
Immunogen Mouse Ig cocktail
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR