Anti-IgG1 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RMG1-1]

Supplier: Biolegend
406613 406614
CA10761-382EA 142.03 CAD
CA10761-382 CA10761-384
Anti-IgG1 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RMG1-1]
Antibodies
PE/Cy7 anti-mouse IgG1 [RMG1-1]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: RMG1-1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR