Anti-IgG Isotype Ctrl Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: HTK888]

Supplier: Biolegend
400943 400944
CA10761-212EA 325.54 CAD
CA10761-212 CA10761-214
Anti-IgG Isotype Ctrl Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: HTK888]
Antibodies
Brilliant Violet 605™ Armenian Hamster IgG Isotype Ctrl [HTK888]; Isotype: Armenian Hamster IgG; Reactivity: Armenian Hamster; Apps: FC, ICFC; Size: 50 μg

Each lot of this Armenian hamster IgG antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG Isotype Ctrl
Clonality: Monoclonal
Clone: HTK888
Conjugation: Brilliant Violet® 605
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Armenian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR