Anti-IgD Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: IA6-2]

Supplier: Biolegend
348227 348228
CA10758-808EA 253.73 CAD
CA10758-808 CA10758-810
Anti-IgD Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: IA6-2]
Antibodies
Alexa Fluor® 647 anti-human IgD [IA6-2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, IHC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgD
Clonality: Monoclonal
Clone: IA6-2
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR