Anti-IgD Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 11-26c.2a]

Supplier: Biolegend
405719 405720
CA10760-992EA 205.86 CAD
CA10760-992 CA10760-994
Anti-IgD Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 11-26c.2a]
Antibodies
PE/Cy7 anti-mouse IgD [11-26c.2a]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgD
Clonality: Monoclonal
Clone: 11-26c.2a
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR