Anti-IgD Mouse Monoclonal Antibody (Biotin) [clone: IA6-2]

Supplier: Biolegend
348211 348212
CA348211-BLEA 113.3 CAD
CA348211-BL CA348212-BL
Anti-IgD Mouse Monoclonal Antibody (Biotin) [clone: IA6-2]
Antibodies
Biotin anti-human IgD [IA6-2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgD
Clonality: Monoclonal
Clone: IA6-2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR