Anti-Ig light chain k Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRK-81]

Supplier: Biolegend
407805 407806
CA10761-418EA 175.54 CAD
CA10761-418 CA10761-420
Anti-Ig light chain k Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRK-81]
Antibodies
PE anti-rat Ig light chain κ [MRK-81]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.03 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ig light chain k
Clonality: Monoclonal
Clone: MRK-81
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR