Anti-IFN-g Rat Monoclonal Antibody [clone: XMG1.2]

Supplier: Biolegend
505833 505834
CA505833-BLEA 175.54 CAD
CA505833-BL CA505834-BL
Anti-IFN-g Rat Monoclonal Antibody [clone: XMG1.2]
Antibodies
Ultra-LEAF™ Purified anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, ELISPOT Capture, CyTOF®, Neut, ICFC, IHC, WB; Size: 1 mg

Each lot of this antibody is quality control tested by ELISA assay. For ELISPOT Capture, the suggested use of this antibody is 1 - 4 μg/ml. For ELISA capture applications, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IFN-γ recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: XMG1.2
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR