Anti-IFN-g Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 4S.B3]
Supplier: Biolegend
502527 502528
CA502527-BLEA 225.01 CAD
CA502527-BL CA502528-BL
Anti-IFN-g Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 4S.B3]
Antibodies
PE/Cy7 anti-human IFN-γ [4S.B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for eachMore Product Information