Anti-IFN-g Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 4S.B3]

Supplier: Biolegend
502521 502522
CA502521-BLEA 197.88 CAD
CA502521-BL CA502522-BL
Anti-IFN-g Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 4S.B3]
Antibodies
Pacific Blue™ anti-human IFN-γ [4S.B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: 4S.B3
Conjugation: PB (Pacific Blue)
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR