Anti-HIF1alpha Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 546-16]

Supplier: Biolegend
359703 359704
CA10760-654EA 244.16 CAD
CA10760-654 CA10760-656
Anti-HIF1alpha Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 546-16]
Antibodies
PE anti-human HIF1α [546-16]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: HIF1alpha
Clonality: Monoclonal
Clone: 546-16
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR