Anti-Granzyme B Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: GB11]

Supplier: Biolegend
515407 515408
CA10761-982EA 280.86 CAD
CA10761-982 CA10761-984
Anti-Granzyme B Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: GB11]
Antibodies
Pacific Blue™ anti-human/mouse Granzyme B [GB11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Cross-Reactivity: Rat; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Granzyme B
Clonality: Monoclonal
Clone: GB11
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse;Cross-Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR