Anti-GM-CSF Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: BVD2-21C11]

Supplier: Biolegend
502313 502314
CA10761-612EA 253.73 CAD
CA10761-612 CA10761-614
Anti-GM-CSF Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: BVD2-21C11]
Antibodies
Pacific Blue™ anti-human GM-CSF [BVD2-21C11]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Rhesus, Cynomolgus; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: BVD2-21C11
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;Cross-Reactivity: Rhesus;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR