Anti-GM-CSF Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: BVD2-21C11]

Supplier: Biolegend
502311 502312
CA502311-BLEA 280.86 CAD
CA502311-BL CA502312-BL
Anti-GM-CSF Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: BVD2-21C11]
Antibodies
PerCP/Cy5.5 anti-human GM-CSF [BVD2-21C11]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Rhesus, Cynomolgus; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: BVD2-21C11
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;Cross-Reactivity: Rhesus;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR