Anti-GL7 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GL7]

Supplier: Biolegend
144613 144614
CA10757-384EA 178.73 CAD
CA10757-384 CA10757-386
Anti-GL7 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GL7]
Antibodies
Pacific Blue™ anti-mouse/human GL7 Antigen (T and B cell Activation Marker) [GL7]; Isotype: Rat IgM, κ; Reactivity: Mouse, Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GL7
Clonality: Monoclonal
Clone: GL7
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR