Anti-GFP Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: FM264G]

Supplier: Biolegend
338007 338008
CA10758-570EA 234.58 CAD
CA10758-570 CA10758-572
Anti-GFP Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: FM264G]
Antibodies
Alexa Fluor® 488 anti-GFP [FM264G]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: FM264G
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR