Anti-Galectin-9 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RG9-35]

Supplier: Biolegend
136109 136110
CA10759-312EA 180.33 CAD
CA10759-312 CA10759-314
Anti-Galectin-9 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RG9-35]
Antibodies
APC anti-mouse Galectin-9 [RG9-35]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Galectin-9
Clonality: Monoclonal
Clone: RG9-35
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR