Anti-Fc epsilon RI alpha Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: AER-37 (CRA-1)]

Supplier: Biolegend
334623 334624
CA10758-540EA 325.54 CAD
CA10758-540 CA10758-542
Anti-Fc epsilon RI alpha Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: AER-37 (CRA-1)]
Antibodies
Brilliant Violet 421™ anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Fc epsilon RI alpha
Clonality: Monoclonal
Clone: AER-37 (CRA-1)
Conjugation: BrilliantMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR