Anti-DYKDDDDK Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: L5]

Supplier: Biolegend
637309 637310
CA10763-226EA 186.71 CAD
CA10763-226 CA10763-228
Anti-DYKDDDDK Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: L5]
Antibodies
PE anti-DYKDDDDK [L5]; Isotype: Rat IgG2a, λ; Reactivity: DYKDDDDK tag epitope; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: L5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, λ
Reactivity: DYKDDDDK tag epitope
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR