Anti-DLL4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MHD4-46]

Supplier: Biolegend
346507 346508
CA346507-BLEA 237.77 CAD
CA346507-BL CA346508-BL
Anti-DLL4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MHD4-46]
Antibodies
APC anti-human Delta-like protein 4 (DLL4) [MHD4-46]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: MHD4-46
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR